missing translation for 'onlineSavingsMsg'
Learn More

N,N,N',N'-Tetraethylethylenediamine 98.0+%, TCI America™

Catalog No. T33715ML
Change view
Click to view available options
Quantity:
5 mL
25 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T33715ML 5 mL
T337125ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T33715ML Supplier TCI America Supplier No. T33715ML

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 150-77-6
Molecular Formula C10H26N2
Molecular Weight (g/mol) 174.33
MDL Number MFCD00009055
InChI Key DIHKMUNUGQVFES-UHFFFAOYSA-P
Synonym n,n,n',n'-tetraethylethylenediamine, teeda, 1,2-ethanediamine, n,n,n',n'-tetraethyl, tetraethylethylenediamine, ethylenediamine, n,n,n',n'-tetraethyl, 1,2-ethanediamine,n1,n1,n2,n2-tetraethyl, n,n,n',n'-tetraethylenediamine, 1,2-ethanediamine, n1,n1,n2,n2-tetraethyl, 2-diethylamino ethyl diethylamine, acmc-20ak9v
PubChem CID 67423
IUPAC Name [2-(diethylazaniumyl)ethyl]diethylazanium
SMILES CC[NH+](CC)CC[NH+](CC)CC

Specifications

Color Yellow
Boiling Point 190°C
Quantity 5 mL
UN Number 2734
Formula Weight 172.32
Percent Purity ≥98.0% (GC,T)
Physical Form Liquid
Chemical Name or Material N,N,N′,N′-Tetraethylethylenediamine