accessibility menu, dialog, popup

UserName

NH2OHO

CAS RN 150-30-1

IUPAC Name: 2-amino-3-phenylpropanoic acid
Synonyms: phenylalanin|fenilalanina|DL-3-phenylalanine|alanine, phenyl-, D-|sabiden|alanine, phenyl-, dl-|phenylalanine DL-form|alanine, phenyl-, DL- (8CI)|polyphenylalanine
Molecular Weight (g/mol): 165.19
Molecular Formula: C9H11NO2
InChi Key: COLNVLDHVKWLRT-UHFFFAOYNA-N
SMILES: NC(CC1=CC=CC=C1)C(O)=O
Percent Purity 
  • (1)
  • (6)
Quantity 
  • (2)
  • (3)
  • (2)
Physical Form 
  • (1)
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (5)
  • (1)
  • (6)
  • (1)
  • (6)
  • (2)
  • (3)
  • (2)
  • (1)
  • (3)