CAS RN 149845-06-7
CAS RN 149845-06-7
IUPAC Name:
(2S)-N-[(2S,3R)-4-[(3S,4aS,8aS)-3-(tert-butylcarbamoyl)-decahydroisoquinolin-2-yl]-3-hydroxy-1-phenylbutan-2-yl]-2-[(quinolin-2-yl)formamido]butanediamide; methanesulfonic acid
Synonyms:
saquinavir
Molecular Weight (g/mol):
766.96
Molecular Formula:
C39H54N6O8S
InChi Key:
IRHXGOXEBNJUSN-YOXDLBRISA-N
SMILES:
CS(O)(=O)=O.CC(C)(C)NC(=O)[C@@H]1C[C@@H]2CCCC[C@@H]2CN1C[C@@H](O)[C@H](CC1=CC=CC=C1)NC(=O)[C@H](CC(N)=O)NC(=O)C1=CC=C2C=CC=CC2=N1
1
–
1
of
1
results
1
Tocris Bioscience™ Saquinavir mesylate
CAS: 149845-06-7 Molecular Formula: C39H54N6O8S