CAS RN 149793-69-1
IUPAC Name:
3-fluoro-5-(trifluoromethyl)benzonitrile
Synonyms:
3-fluoro-5-(trifluoromethyl)benzonitrile
Molecular Weight (g/mol):
189.11
Molecular Formula:
C8H3F4N
InChi Key:
AYNUKEDZAYOEGG-UHFFFAOYSA-N
SMILES:
FC1=CC(=CC(=C1)C(F)(F)F)C#N
1
–
1
of
1
results
1
3-Fluoro-5-(trifluoromethyl)benzonitrile 98.0+%, TCI America™
CAS: 149793-69-1 Molecular Formula: C8H3F4N Molecular Weight (g/mol): 189.113 MDL Number: MFCD00061282 InChI Key: AYNUKEDZAYOEGG-UHFFFAOYSA-N Synonym: 3-fluoro-5-trifluoromethyl benzonitrile,3-cyano-5-fluorobenzotrifluoride,3-fluoro-5-trifluoromethyl-benzonitrile,3-fluoro-5 trifluoromethyl benzonitrile,benzonitrile, 3-fluoro-5-trifluoromethyl,alpha,alpha,alpha,5-tetrafluoro-m-tolunitrile,3-fluoro-5-trifluoromethyl benzenecarbonitrile,.alpha.,.alpha.,.alpha.,5-tetrafluoro-m-tolunitrile,pubchem2620 PubChem CID: 519037 IUPAC Name: 3-fluoro-5-(trifluoromethyl)benzonitrile SMILES: C1=C(C=C(C=C1C(F)(F)F)F)C#N