accessibility menu, dialog, popup

UserName

CAS RN 149104-88-1

IUPAC Name: (4-methanesulfonylphenyl)boronic acid
Synonyms: 4-methanesulfonylphenylboronic acid
Molecular Weight (g/mol): 200.02
Molecular Formula: C7H9BO4S
InChi Key: VDUKDQTYMWUSAC-UHFFFAOYSA-N
SMILES: CS(=O)(=O)C1=CC=C(C=C1)B(O)O
Physical Form 
  • (1)
  • (2)
Quantity 
  • (2)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (2)
  • (1)
  • (2)
  • (2)
  • (1)