accessibility menu, dialog, popup

UserName

SeSe

CAS RN 1482-82-2

IUPAC Name: [(benzyldiselanyl)methyl]benzene
Synonyms: [(benzyldiselanyl)methyl]benzene
Molecular Weight (g/mol): 340.21
Molecular Formula: C14H14Se2
InChi Key: HYAVEDMFTNAZQE-UHFFFAOYSA-N
SMILES: C([Se][Se]CC1=CC=CC=C1)C1=CC=CC=C1
Percent Purity 
  • (1)
  • (3)
Quantity 
  • (1)
  • (1)
  • (2)
Physical Form 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (1)
  • (3)
  • (1)
  • (3)
  • (1)
  • (1)
  • (2)
  • (1)