missing translation for 'onlineSavingsMsg'
Learn More

Dibenzyl Diselenide 95.0+%, TCI America™

Catalog No. D27175G
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D27175G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D27175G Supplier TCI America Supplier No. D27175G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 1482-82-2
Molecular Formula C14H14Se2
Molecular Weight (g/mol) 340.208
MDL Number MFCD00004767
InChI Key HYAVEDMFTNAZQE-UHFFFAOYSA-N
Synonym dibenzyl diselenide, 1,2-dibenzyldiselane, benzyl diselenide, dibenzyldiselenide, dibenzyldiselane, diselenide, bis phenylmethyl, dibenzyldiselenium, bisbenzyldiselenide, diselenide,bis phenylmethyl, benzyldiselanyl methylbenzene
PubChem CID 95955
IUPAC Name (benzyldiselanyl)methylbenzene
SMILES C1=CC=C(C=C1)C[Se][Se]CC2=CC=CC=C2

Specifications

Melting Point 93°C
Color Yellow
Quantity 5 g
UN Number 3283
Formula Weight 340.19
Percent Purity ≥95.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Dibenzyl Diselenide