missing translation for 'onlineSavingsMsg'
Learn More

Dibenzyl diselenide, 95%

Catalog No. AAB2127814
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2127814 25 g
AAB2127803 1 g
AAB2127806 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAB2127814 Supplier Thermo Scientific Chemicals Supplier No. B2127814
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 1482-82-2 | C14H14Se2 | 340.208 g/mol

Dibenzyl diselenide is used as a source of the PhSe unit in organic synthesis. It is also used to introduce PhSe groups by reaction with a variety of nucleophilic, including enolates, enol silyl ethers, Grignard reagents, organolithium reagents, alkenes and amines.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Dibenzyl diselenide is used as a source of the PhSe unit in organic synthesis. It is also used to introduce PhSe groups by reaction with a variety of nucleophilic, including enolates, enol silyl ethers, Grignard reagents, organolithium reagents, alkenes and amines.

Solubility
Soluble in water.

Notes
Air Sensitive. Keep container tightly closed. Store away from oxidizing agents.

Chemical Identifiers

CAS 1482-82-2
Molecular Formula C14H14Se2
Molecular Weight (g/mol) 340.208
MDL Number MFCD00004767
InChI Key HYAVEDMFTNAZQE-UHFFFAOYSA-N
Synonym dibenzyl diselenide, 1,2-dibenzyldiselane, benzyl diselenide, dibenzyldiselenide, dibenzyldiselane, diselenide, bis phenylmethyl, dibenzyldiselenium, bisbenzyldiselenide, diselenide,bis phenylmethyl, benzyldiselanyl methylbenzene
PubChem CID 95955
IUPAC Name (benzyldiselanyl)methylbenzene
SMILES C1=CC=C(C=C1)C[Se][Se]CC2=CC=CC=C2

Specifications

Melting Point 91°C to 93°C
Quantity 25 g
UN Number UN3283
Beilstein 1877745
Solubility Information Soluble in water.
Formula Weight 340.18
Percent Purity 95%
Chemical Name or Material Dibenzyl diselenide
Hazard Category H301+H331-H373
Hazard Statement GHS H Statement
H301-H331-H373
Toxic if swallowed.
Toxic if inhaled.
May cause damage to organs through prolonged or repeated exposure.
Precautionary Statement P260-P264b-P270-P271-P301+P310-P304+P340-P311-P314-P330-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: SELENIUM COMPOUND, SOLID, N.O.S.
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only