accessibility menu, dialog, popup

UserName

CAS RN 148043-73-6

IUPAC Name: 4,4,5,5,5-pentafluoropentan-1-ol
Synonyms: 4,4,5,5,5-pentafluoropentan-1-ol
Molecular Weight (g/mol): 178.1
Molecular Formula: C5H7F5O
InChi Key: QROUUECTKRZFHF-UHFFFAOYSA-N
SMILES: OCCCC(F)(F)C(F)(F)F
Boiling Point 
  • (2)
  • (4)
Molecular Weight (g/mol) 
  • (2)
  • (4)
Percent Purity 
  • (2)
  • (2)
  • (2)
Quantity 
  • (1)
  • (1)
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (4)
  • (2)
  • (4)
  • (2)
  • (4)
  • (2)
  • (4)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)
  • (2)
  • (2)
  • (4)