missing translation for 'onlineSavingsMsg'
Learn More

4,4,5,5,5-Pentafluoro-1-pentanol 93.0+%, TCI America™

Catalog No. P13425G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P13425G 5 g
P134225G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. P13425G Supplier TCI America Supplier No. P13425G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 148043-73-6
Molecular Formula C5H7F5O
Molecular Weight (g/mol) 178.102
MDL Number MFCD00153224
InChI Key QROUUECTKRZFHF-UHFFFAOYSA-N
Synonym 4,4,5,5,5-pentafluoro-1-pentanol, 4,4,5,5,5-pentafluoropentanol, pentafluoropentanol, 1-pentanol, 4,4,5,5,5-pentafluoro, fulvestrant intermediate, acmc-1bz8x, 3-perfluoroethyl propan-1-ol, 4,4,5,5,5-pentafluoro pentanol, 4,4,5,5,5-pentafluoro-pentanol, 4,4,5,5,5-pentafluorpentan-1-ol
PubChem CID 547967
IUPAC Name 4,4,5,5,5-pentafluoropentan-1-ol
SMILES C(CC(C(F)(F)F)(F)F)CO

Specifications

Color Yellow
Boiling Point 135°C
Quantity 5 g
Formula Weight 178.10
Percent Purity ≥93.0% (GC)
Physical Form Liquid
Chemical Name or Material 4,4,5,5,5-Pentafluoro-1-pentanol
EINECSNumber (2)-3955
TSCA No