accessibility menu, dialog, popup

UserName

H3CSiCH3ClCl

CAS RN 14799-93-0

IUPAC Name: dichloro(methyl)octylsilane
Synonyms: dichloro(methyl)octylsilane
Molecular Weight (g/mol): 227.24
Molecular Formula: C9H20Cl2Si
InChi Key: QHBMMABVNRSRHW-UHFFFAOYSA-N
SMILES: CCCCCCCC[Si](C)(Cl)Cl

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results