missing translation for 'onlineSavingsMsg'
Learn More

Dichloro(methyl)-n-octylsilane 97.0+%, TCI America™

Catalog No. O026525ML
Change view
Click to view available options
Quantity:
25 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
O026525ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. O026525ML Supplier TCI America Supplier No. O026525ML

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 14799-93-0
Molecular Formula C9H20Cl2Si
Molecular Weight (g/mol) 227.24
MDL Number MFCD00042606
InChI Key QHBMMABVNRSRHW-UHFFFAOYSA-N
Synonym n-octylmethyldichlorosilane, silane, dichloromethyloctyl, dichloro methyl-n-octylsilane, methyloctyldichlorosilane, dichloro methyl octyl silane, dichloro methyl octylsilane, octylmethyldichlorosilane, acmc-209czn, methyloctyl dichlorosilane, dichloro methyl octylsilane #
PubChem CID 84649
IUPAC Name dichloro(methyl)octylsilane
SMILES CCCCCCCC[Si](C)(Cl)Cl

Specifications

Boiling Point 234°C
Quantity 25 mL
UN Number 2986
Formula Weight 227.24
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material Dichloro(methyl)-n-octylsilane
EINECSNumber (2)-2041&(2)-2047
TSCA Yes