accessibility menu, dialog, popup

UserName

H3CONOO(E)

CAS RN 1472-68-0

IUPAC Name: 1-methoxy-4-[(1E)-2-(4-nitrophenyl)ethenyl]benzene
Synonyms: (E)-methoxy-4'-nitrostilbene
Molecular Weight (g/mol): 255.27
Molecular Formula: C15H13NO3
InChi Key: PDFBJCRLHMEJQP-NSCUHMNNSA-N
SMILES: COC1=CC=C(\C=C\C2=CC=C(C=C2)[N+]([O-])=O)C=C1
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results