accessibility menu, dialog, popup

UserName

(S)(R)(S)O(S)(R)OOHHOHOHO

CAS RN 1464-44-4

IUPAC Name: (2R,3S,4S,5R,6S)-2-(hydroxymethyl)-6-phenoxyoxane-3,4,5-triol
Synonyms: phenyl β-D-glucoside
Molecular Weight (g/mol): 256.25
Molecular Formula: C12H16O6
InChi Key: NEZJDVYDSZTRFS-WLEIBRHLNA-N
SMILES: OC[C@H]1O[C@@H](OC2=CC=CC=C2)[C@H](O)[C@@H](O)[C@@H]1O
Percent Purity 
  • (1)
  • (2)
Physical Form 
  • (1)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)