accessibility menu, dialog, popup

UserName

CH3

CAS RN 1459-55-8

IUPAC Name: 1-(4-methylphenyl)adamantane
Synonyms: 1-(4-methylphenyl)adamantane
Molecular Weight (g/mol): 226.36
Molecular Formula: C17H22
InChi Key: RFQXTZGKFOTWFL-UHFFFAOYSA-N
SMILES: CC1=CC=C(C=C1)C12CC3CC(CC(C3)C1)C2
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results