missing translation for 'onlineSavingsMsg'
Learn More

p-(1-Adamantyl)toluene 95.0+%, TCI America™

Catalog No. A13935G
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A13935G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. A13935G Supplier TCI America Supplier No. A13935G
Only null left

Chemical Identifiers

CAS 1459-55-8
Molecular Formula C17H22
Molecular Weight (g/mol) 226.363
MDL Number MFCD00181038
InChI Key RFQXTZGKFOTWFL-UHFFFAOYSA-N
Synonym 1-(4-Methylphenyl)adamantane, 1-(p-Tolyl)adamantane
PubChem CID 300774
IUPAC Name 1-(4-methylphenyl)adamantane
SMILES CC1=CC=C(C=C1)C23CC4CC(C2)CC(C4)C3

Specifications

Melting Point 99°C
Color White-Yellow
Quantity 5 g
Formula Weight 226.36
Percent Purity ≥95.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material p-(1-Adamantyl)toluene