accessibility menu, dialog, popup

UserName

CH3BOHHOCl

CAS RN 145349-62-8

IUPAC Name: (2-chloro-4-methylphenyl)boronic acid
Synonyms: 2-chloro-4-methylphenylboronic acid
Molecular Weight (g/mol): 170.4
Molecular Formula: C7H8BClO2
InChi Key: UKYCKUPXBPLXBA-UHFFFAOYSA-N
SMILES: CC1=CC=C(B(O)O)C(Cl)=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (2)
  • (2)
  • (1)