missing translation for 'onlineSavingsMsg'
Learn More

2-Chloro-4-methylbenzeneboronic acid, 95%

Catalog No. AAH2860406
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH2860406 5 g
AAH2860403 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH2860406 Supplier Thermo Scientific Chemicals Supplier No. H2860406
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 145349-62-8 | C7H8BClO2 | 170.399 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 145349-62-8
Molecular Formula C7H8BClO2
Molecular Weight (g/mol) 170.399
MDL Number MFCD03411936
InChI Key UKYCKUPXBPLXBA-UHFFFAOYSA-N
Synonym 2-chloro-4-methylphenyl boronic acid, 2-chloro-4-methylbenzeneboronic acid, boronic acid, 2-chloro-4-methylphenyl, pubchem1778, 2-chloro-4-methyl-phenyl boronic acid, acmc-209cuq, ksc524i9h, 2-chloro-p-tolylboronic acid, 2-chloro-4-methyl phenyl boronic acid
PubChem CID 2773334
IUPAC Name (2-chloro-4-methylphenyl)boronic acid
SMILES B(C1=C(C=C(C=C1)C)Cl)(O)O

Specifications

Melting Point 198°C to 202°C
Quantity 5 g
Formula Weight 170.4
Percent Purity 97%
Chemical Name or Material 2-Chloro-4-methylbenzeneboronic acid
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only