accessibility menu, dialog, popup

UserName

CH3OOONOOCH3H3CCH3

CAS RN 144978-12-1

IUPAC Name: 1-tert-butyl 2-ethyl 5-oxopyrrolidine-1,2-dicarboxylate
Synonyms: 1-tert-butyl 2-ethyl 5-oxopyrrolidine-1,2-dicarboxylate
Molecular Weight (g/mol): 257.29
Molecular Formula: C12H19NO5
InChi Key: YWWWGFSJHCFVOW-UHFFFAOYNA-N
SMILES: CCOC(=O)C1CCC(=O)N1C(=O)OC(C)(C)C
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (2)
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)