accessibility menu, dialog, popup

UserName

CAS RN 1443438-29-6

NSNHNOHO

CAS RN 1443438-29-6

IUPAC Name: 1-[(pyridin-3-yl)carbamothioyl]pyrrolidine-2-carboxylic acid
Synonyms: 1-[(pyridin-3-yl)carbamothioyl]pyrrolidine-2-carboxylic acid
Molecular Weight (g/mol): 251.3
Molecular Formula: C11H13N3O2S
InChi Key: WDIJWXTYNMSAKY-UHFFFAOYNA-N
SMILES: OC(=O)C1CCCN1C(=S)NC1=CN=CC=C1
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results