missing translation for 'onlineSavingsMsg'
Learn More

(R)-1-(3-Pyridylthiocarbamoyl)pyrrolidine-2-carboxylic Acid 98.0+%, TCI America™

Catalog No. P2298100MG
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P2298100MG 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. P2298100MG Supplier TCI America Supplier No. P2298100MG
Only null left

Chemical Identifiers

CAS 1443438-29-6
Molecular Formula C11H13N3O2S
Molecular Weight (g/mol) 251.304
InChI Key WDIJWXTYNMSAKY-VIFPVBQESA-N
Synonym (R)-PyT-C
PubChem CID 102443407
IUPAC Name (2S)-1-(pyridin-3-ylcarbamothioyl)pyrrolidine-2-carboxylic acid
SMILES C1CC(N(C1)C(=S)NC2=CN=CC=C2)C(=O)O

Specifications

Melting Point 192°C
Color White
Quantity 100 mg
Formula Weight 251.30
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material (R)-1-(3-Pyridylthiocarbamoyl)pyrrolidine-2-carboxylic Acid