accessibility menu, dialog, popup

UserName

CH3SNHSOOH2NNN

CAS RN 144-82-1

IUPAC Name: 4-amino-N-(5-methyl-1,3,4-thiadiazol-2-yl)benzene-1-sulfonamide
Synonyms: sulfamethizole proklar microsul Uroz ultrasul thiosulfil sulfa gram sulfurine sulfamethazole rufol
Molecular Weight (g/mol): 270.33
Molecular Formula: C9H10N4O2S2
InChi Key: VACCAVUAMIDAGB-UHFFFAOYSA-N
SMILES: CC1=NN=C(NS(=O)(=O)C2=CC=C(N)C=C2)S1
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results