missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sulfamethizole, ≥99% (HPLC), MilliporeSigma™ Supelco™
Description
Environmental and Pharmaceutical (small molecule)
Specifications
Specifications
| Packaging | Glass Bottle |
| CAS | 144-82-1 |
| Physical Form | Neat |
| Linear Formula | C9H10N4O2S2 |
| MDL Number | MFCD00053363 |
| Synonym | 4-Amino-N-(5-methyl-1,3,4-thiadiazol-2-yl)benzenesulfonamide; N1-(5-Methyl-1,3,4-thiadiazol-2-yl)sulfanilamide; Sulfamethylthiadiazole |
| Type | Analytical Standard |
| Quantity | 25 g |
| Formula Weight | 270.33 |
| Percent Purity | ≥99% (HPLC) |
| Show More |