accessibility menu, dialog, popup

UserName

CH3SiCl

CAS RN 144-79-6

IUPAC Name: chloro(methyl)diphenylsilane
Synonyms: silane, chloromethyldiphenyl-
Molecular Weight (g/mol): 232.78
Molecular Formula: C13H13ClSi
InChi Key: OJZNZOXALZKPEA-UHFFFAOYSA-N
SMILES: C[Si](Cl)(C1=CC=CC=C1)C1=CC=CC=C1
Percent Purity 
  • (1)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results