missing translation for 'onlineSavingsMsg'
Learn More

Chloro(methyl)diphenylsilane, 97%

Catalog No. AAL0416218
Change view
Click to view available options
Quantity:
10 g
50 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL0416218 50 g
AAL0416209 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL0416218 Supplier Thermo Scientific Chemicals Supplier No. L0416218
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 144-79-6 | C13H13ClSi | 232.78 g/mol

Chloro(methyl)diphenylsilane is used in agrochemical, pharmaceutical and dyestuff field .

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Chloro(methyl)diphenylsilane is used in agrochemical, pharmaceutical and dyestuff field .

Solubility
Reacts with water.

Notes
Moisture Sensitive. Store away from strong bases. Incompatible with moisture and bases.

Chemical Identifiers

CAS 144-79-6
Molecular Formula C13H13ClSi
Molecular Weight (g/mol) 232.78
MDL Number MFCD00000498
InChI Key OJZNZOXALZKPEA-UHFFFAOYSA-N
Synonym chloro methyl diphenylsilane, methyldiphenylchlorosilane, chloromethyldiphenylsilane, silane, chloromethyldiphenyl, chlorodiphenylmethylsilane, diphenylmethylchlorosilane, methyldiphenylsilyl chloride, dpmscl, chloro-methyl-diphenyl-silane, benzene, 1,1'-chloromethylsilylene bis
PubChem CID 67349
IUPAC Name chloro-methyl-diphenylsilane
SMILES C[Si](Cl)(C1=CC=CC=C1)C1=CC=CC=C1

Specifications

Boiling Point 294°C to 295°C
Quantity 50 g
UN Number UN2987
Beilstein 2937445
Flash Point 141°C (285°F)
Solubility Information Reacts with water.
Formula Weight 232.79
Percent Purity 97%
Odor Irritating
Refractive Index 1.574
Sensitivity Moisture sensitive
Density 1.104
Chemical Name or Material Chloro(methyl)diphenylsilane
Show More Show Less
Hazard Category H314
Hazard Statement GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
Precautionary Statement P260-P264b-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
DOTInformation Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CHLOROSILANES, CORROSIVE, N.O.S.
EINECSNumber 205-639-0
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only