missing translation for 'onlineSavingsMsg'
Learn More

Diphenylmethylchlorosilane 95.0+%, TCI America™

Catalog No. D139025ML
Change view
Click to view available options
Quantity:
25 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D139025ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D139025ML Supplier TCI America Supplier No. D139025ML

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 144-79-6
Molecular Formula C13H13ClSi
Molecular Weight (g/mol) 232.78
MDL Number MFCD00000498
InChI Key OJZNZOXALZKPEA-UHFFFAOYSA-N
Synonym chloro methyl diphenylsilane, methyldiphenylchlorosilane, chloromethyldiphenylsilane, silane, chloromethyldiphenyl, chlorodiphenylmethylsilane, diphenylmethylchlorosilane, methyldiphenylsilyl chloride, dpmscl, chloro-methyl-diphenyl-silane, benzene, 1,1'-chloromethylsilylene bis
PubChem CID 67349
IUPAC Name chloro(methyl)diphenylsilane
SMILES C[Si](Cl)(C1=CC=CC=C1)C1=CC=CC=C1

Specifications

Melting Point -22°C
Color Yellow
Boiling Point 295°C
Quantity 25 mL
UN Number 2987
Formula Weight 232.78
Percent Purity ≥95.0% (GC)
Physical Form Liquid
Chemical Name or Material Diphenylmethylchlorosilane
EINECSNumber (3)-2641
TSCA Yes