CAS RN 14362-31-3
CAS RN 14362-31-3
IUPAC Name:
hydrogen 1-[(4-chlorophenyl)(phenyl)methyl]-4-methylpiperazine chloride
Synonyms:
hydrogen chlorcyclizine chloride
Molecular Weight (g/mol):
337.29
Molecular Formula:
C18H22Cl2N2
InChi Key:
MSIJLVMSKDXAQN-UHFFFAOYNA-N
SMILES:
[H+].[Cl-].CN1CCN(CC1)C(C1=CC=CC=C1)C1=CC=C(Cl)C=C1
1
–
1
of
1
results
1
Chlorcyclizine Hydrochloride, British Pharmacopoeia (BP) Reference Standard, MilliporeSigma™ Supelco™
This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets, has been developed and issued under the Authority of the issuing Pharmacopoeia.