missing translation for 'onlineSavingsMsg'
Learn More

Chlorcyclizine Hydrochloride, British Pharmacopoeia (BP) Reference Standard, MilliporeSigma™ Supelco™

Catalog No. 111022833
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111022833 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111022833 Supplier Merck Emd Millipore Supplier No. BP067

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets, has been developed and issued under the Authority of the issuing Pharmacopoeia.

Specifications

Chemical Name or Material Pharmacopeia & Metrological Institutes Standards
CAS 14362-31-3
Molecular Formula C18H21ClN2 · HCl
Linear Formula C18H21ClN2 · HCl
Quantity 100 mg
Synonym 1-[(4-Chlorophenyl)phenylmethyl]-4-methylpiperazine monohydrochloride
Formula Weight 337.29
Grade British Pharmacopoeia (BP) Reference Standard