accessibility menu, dialog, popup

UserName

CAS RN 1431-37-4

H3COOOOCH3

CAS RN 1431-37-4

IUPAC Name: 1,3-dihexyl propanedioate
Synonyms: 1,3-dihexyl propanedioate
Molecular Weight (g/mol): 272.39
Molecular Formula: C15H28O4
InChi Key: MQXAJNXSULJYCY-UHFFFAOYSA-N
SMILES: CCCCCCOC(=O)CC(=O)OCCCCCC

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results