missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Dihexyl Malonate 98.0+%, TCI America™
Specifications
Specifications
| Color | Colorless |
| Quantity | 5 g |
| Formula Weight | 272.39 |
| Percent Purity | ≥98.0% (GC) |
| Physical Form | Liquid |
| Chemical Name or Material | Dihexyl Malonate |
Safety and Handling
| TSCA | No |