accessibility menu, dialog, popup

UserName

H2CFF

CAS RN 142400-92-8

IUPAC Name: 4-(3,4-difluorophenyl)-4'-ethenyl-1,1'-bi(cyclohexane)
Synonyms: 4-(3,4-difluorophenyl)-4'-ethenyl-1,1'-bi(cyclohexane)
Molecular Weight (g/mol): 304.43
Molecular Formula: C20H26F2
InChi Key: ALFLDQIYGBNZCO-UHFFFAOYSA-N
SMILES: FC1=C(F)C=C(C=C1)C1CCC(CC1)C1CCC(CC1)C=C
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results