accessibility menu, dialog, popup

UserName

NOOHH2NONHOHNOH2N

CAS RN 141801-26-5

IUPAC Name: 2-({1-[2-amino-3-(4-hydroxyphenyl)propanoyl]pyrrolidin-2-yl}formamido)-N-(1-carbamoyl-2-phenylethyl)-3-phenylpropanamide
Synonyms: endomorphin-2
Molecular Weight (g/mol): 571.68
Molecular Formula: C32H37N5O5
InChi Key: XIJHWXXXIMEHKW-UHFFFAOYNA-N
SMILES: NC(CC1=CC=C(O)C=C1)C(=O)N1CCCC1C(=O)NC(CC1=CC=CC=C1)C(=O)NC(CC1=CC=CC=C1)C(N)=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)