missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Endomorphin-2

Catalog No. AAJ66953MB
Change view
Click to view available options
Quantity:
5mg
25mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ66953MB 25mg
AAJ66953LB0 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAJ66953MB Supplier Thermo Scientific Chemicals Supplier No. J66953MB

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 141801-26-5

Specifications

Color White
Content And Storage Store at -20°C
Format Solid
Molecular Weight (g/mol) 571.678
Product Type Endomorphin-2
Quantity 25mg

RUO – Research Use Only