missing translation for 'onlineSavingsMsg'
Learn More

4-Bromo-DL-phenylalanine 98.0+%, TCI America™

Catalog No. B42455G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B42455G 5 g
B42451G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B42455G Supplier TCI America Supplier No. B42455G

Chemical Identifiers

CAS 14091-15-7
Molecular Formula C9H10BrNO2
Molecular Weight (g/mol) 244.09
MDL Number MFCD00002599
InChI Key PEMUHKUIQHFMTH-UHFFFAOYSA-N
Synonym 2-amino-3-4-bromophenyl propanoic acid, p-bromo-dl-phenylalanine, 4-bromo-dl-phenylalanine, dl-4-br-phe-oh, 4-bromophenylalanine, 4-bromo-phenylalanine, h-dl-phe 4-br-oh, 2-amino-3-4-bromophenyl propionic acid, 2-amino-3-4-bromo-phenyl-propionic acid, p-br-phenylalanine
PubChem CID 85681
IUPAC Name 2-amino-3-(4-bromophenyl)propanoic acid
SMILES NC(CC1=CC=C(Br)C=C1)C(O)=O

Specifications

Melting Point 258°C
Color Yellow
Quantity 5 g
Formula Weight 244.09
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material 4-Bromo-DL-phenylalanine
TSCA No
Recommended Storage Refrigerator