accessibility menu, dialog, popup

UserName

CAS RN 140-95-4

IUPAC Name: 1,3-bis(hydroxymethyl)urea
Synonyms: dimethylol urea
Molecular Weight (g/mol): 120.11
Molecular Formula: C3H8N2O3
InChi Key: QUBQYFYWUJJAAK-UHFFFAOYSA-N
SMILES: OCNC(=O)NCO
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (1)
  • (1)
  • (2)
Grade 
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)
  • (2)
  • (2)
  • (2)