missing translation for 'onlineSavingsMsg'
Learn More

1,3-Bis(hydroxymethyl)urea, tech. 90%

Catalog No. AAA1606036
Change view
Click to view available options
Quantity:
100 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1606036 500 g
AAA1606022 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1606036 Supplier Thermo Scientific Chemicals Supplier No. A1606036
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 140-95-4 | C3H8N2O3 | 120.11 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 140-95-4
Molecular Formula C3H8N2O3
Molecular Weight (g/mol) 120.11
MDL Number MFCD00014414
InChI Key QUBQYFYWUJJAAK-UHFFFAOYSA-N
Synonym 1,3-bis hydroxymethyl urea, dimethylolurea, oxymethurea, n,n'-dimethylolurea, methural, carbamol, caurite, metural, urofix, protesine dmu
PubChem CID 8827
IUPAC Name 1,3-bis(hydroxymethyl)urea
SMILES OCNC(=O)NCO

Specifications

Melting Point 124°C to 128°C
pH 7 to 8
Odor Odorless
Linear Formula (HOCH2NH)2C=O
Quantity 500 g
Formula Weight 120.11
Percent Purity 90%
Grade Technical
Chemical Name or Material 1,3-Bis(hydroxymethyl)urea
EINECSNumber 205-444-0
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only