missing translation for 'onlineSavingsMsg'
Learn More

1,3-Bis(hydroxymethyl)urea 97.0+%, TCI America™

Catalog No. D0767500G
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D0767500G 500 g
D076725G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D0767500G Supplier TCI America Supplier No. D0767500G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 140-95-4
Molecular Formula C3H8N2O3
Molecular Weight (g/mol) 120.11
MDL Number MFCD00014414
InChI Key QUBQYFYWUJJAAK-UHFFFAOYSA-N
Synonym 1,3-bis hydroxymethyl urea, dimethylolurea, oxymethurea, n,n'-dimethylolurea, methural, carbamol, caurite, metural, urofix, protesine dmu
PubChem CID 8827
IUPAC Name 1,3-bis(hydroxymethyl)urea
SMILES OCNC(=O)NCO

Specifications

Melting Point 133°C
Color White
Quantity 500 g
Formula Weight 120.11
Percent Purity ≥97.0% (N)
Physical Form Crystalline Powder
Chemical Name or Material 1,3-Bis(hydroxymethyl)urea
EINECSNumber (2)-1739
RTECSNumber YS2716350
TSCA Yes
Recommended Storage Refrigerator