accessibility menu, dialog, popup

UserName

ClOO

CAS RN 140-18-1

IUPAC Name: benzyl 2-chloroacetate
Synonyms: benzyl 2-chloroacetate
Molecular Weight (g/mol): 184.62
Molecular Formula: C9H9ClO2
InChi Key: SOGXBRHOWDEKQB-UHFFFAOYSA-N
SMILES: ClCC(=O)OCC1=CC=CC=C1
Boiling Point 
  • (2)
  • (2)
Molecular Weight (g/mol) 
  • (2)
  • (2)
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)