CAS RN 1397202-77-5
CAS RN 1397202-77-5
IUPAC Name:
N,N-diphenylhexacyclo[12.12.0.0²,⁷.0⁸,¹³.0¹⁵,²⁰.0²¹,²⁶]hexacosa-1(14),2,4,6,8,10,12,15,17,19,21,23,25-tridecaen-5-amine
Synonyms:
N,N-diphenylhexacyclo[12.12.0.0²,⁷.0⁸,¹³.0¹⁵,²⁰.0²¹,²⁶]hexacosa-1(14),2,4,6,8,10,12,15,17,19,21,23,25-tridecaen-5-amine
Molecular Weight (g/mol):
495.63
Molecular Formula:
C38H25N
InChi Key:
UNHRFJSSMJBBLW-UHFFFAOYSA-N
SMILES:
C1=CC=C(C=C1)N(C1=CC=CC=C1)C1=CC=C2C(=C1)C1=CC=CC=C1C1=C2C2=CC=CC=C2C2=CC=CC=C12
1
–
1
of
1
results
1
3-(Diphenylamino)dibenzo[g,p]chrysene 97.0+%, TCI America™
CAS: 1397202-77-5 Molecular Formula: C38H25N Molecular Weight (g/mol): 495.625 InChI Key: UNHRFJSSMJBBLW-UHFFFAOYSA-N Synonym: 3-DPADBC PubChem CID: 44629943 SMILES: C1=CC=C(C=C1)N(C2=CC=CC=C2)C3=CC4=C(C=C3)C5=C(C6=CC=CC=C6C7=CC=CC=C75)C8=CC=CC=C84