missing translation for 'onlineSavingsMsg'
Learn More

3-(Diphenylamino)dibenzo[g,p]chrysene 97.0+%, TCI America™

Catalog No. D3739200MG
Change view
Click to view available options
Quantity:
200 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D3739200MG 200 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D3739200MG Supplier TCI America Supplier No. D3739200MG
Only null left

Chemical Identifiers

CAS 1397202-77-5
Molecular Formula C38H25N
Molecular Weight (g/mol) 495.625
InChI Key UNHRFJSSMJBBLW-UHFFFAOYSA-N
Synonym 3-DPADBC
PubChem CID 44629943
SMILES C1=CC=C(C=C1)N(C2=CC=CC=C2)C3=CC4=C(C=C3)C5=C(C6=CC=CC=C6C7=CC=CC=C75)C8=CC=CC=C84

Specifications

Melting Point 234°C
Quantity 200 mg
Formula Weight 495.63
Percent Purity ≥97.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material 3-(Diphenylamino)dibenzo[g,p]chrysene