CAS RN 13957-31-8
IUPAC Name:
1-[(2R,3R,4S,5R)-3,4-dihydroxy-5-(hydroxymethyl)oxolan-2-yl]-4-sulfanylidene-1,2,3,4-tetrahydropyrimidin-2-one
Synonyms:
thiouridine s4u
Molecular Weight (g/mol):
260.26
Molecular Formula:
C9H12N2O5S
InChi Key:
ZLOIGESWDJYCTF-XVFCMESISA-N
SMILES:
OC[C@H]1O[C@H]([C@H](O)[C@@H]1O)N1C=CC(=S)NC1=O
1
–
2
of
2
results
1
4-Thiouridine, 98+%
CAS: 13957-31-8 Molecular Formula: C9H12N2O5S Molecular Weight (g/mol): 260.26 MDL Number: MFCD00006538 InChI Key: ZLOIGESWDJYCTF-XVFCMESISA-N PubChem CID: 3032615 ChEBI: CHEBI:20480 SMILES: OC[C@H]1O[C@H]([C@H](O)[C@@H]1O)N1C=CC(=S)NC1=O
2
R&D Systems™ 4-Thiouridine
A Tocris product, part of R&D Systems. Photoreactive uridine analog for RNA sequencing