accessibility menu, dialog, popup

UserName

H3COBrN

CAS RN 138642-47-4

IUPAC Name: 2-bromo-5-methoxybenzonitrile
Synonyms: 2-bromo-5-methoxybenzonitrile
Molecular Weight (g/mol): 212.05
Molecular Formula: C8H6BrNO
InChi Key: ZQONVYONOASKIY-UHFFFAOYSA-N
SMILES: COC1=CC(C#N)=C(Br)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results