accessibility menu, dialog, popup

UserName

CH2H2NHCl

CAS RN 138286-02-9

IUPAC Name: 2-(prop-2-en-1-yl)aniline hydrochloride
Synonyms: 2-(prop-2-en-1-yl)aniline hydrochloride
Molecular Weight (g/mol): 169.65
Molecular Formula: C9H12ClN
InChi Key: QEUTZGMYXDQKNU-UHFFFAOYSA-N
SMILES: Cl.NC1=CC=CC=C1CC=C
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results