accessibility menu, dialog, popup

UserName

CAS RN 13811-71-7

IUPAC Name: 1,4-diethyl (2S,3S)-2,3-dihydroxybutanedioate
Synonyms: diethyl tartrate
Molecular Weight (g/mol): 206.19
Molecular Formula: C8H14O6
InChi Key: YSAVZVORKRDODB-WDSKDSINSA-N
SMILES: CCOC(=O)[C@@H](O)[C@H](O)C(=O)OCC
Boiling Point 
  • (2)
  • (3)
  • (1)
Percent Purity 
  • (2)
  • (4)
Quantity 
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
Physical Form 
  • (2)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (2)
  • (4)
  • (2)
  • (3)
  • (1)
  • (2)
  • (4)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)