accessibility menu, dialog, popup

UserName

CAS RN 1380288-87-8

H3CCH3NONNNNH2NHOOHNHNCH3CH3H3C

CAS RN 1380288-87-8

IUPAC Name: 2-(6-amino-9H-purin-9-yl)-5-[({3-[2-(6-tert-butyl-1H-1,3-benzodiazol-2-yl)ethyl]cyclobutyl}(propan-2-yl)amino)methyl]oxolane-3,4-diol
Synonyms: 2-(6-aminopurin-9-yl)-5-[({3-[2-(5-tert-butyl-3H-1,3-benzodiazol-2-yl)ethyl]cyclobutyl}(isopropyl)amino)methyl]oxolane-3,4-diol
Molecular Weight (g/mol): 562.72
Molecular Formula: C30H42N8O3
InChi Key: LXFOLMYKSYSZQS-UHFFFAOYNA-N
SMILES: CC(C)N(CC1OC(C(O)C1O)N1C=NC2=C(N)N=CN=C12)C1CC(CCC2=NC3=CC=C(C=C3N2)C(C)(C)C)C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results