accessibility menu, dialog, popup

UserName

OHHOHOOHO

CAS RN 138-59-0

IUPAC Name: 3,4,5-trihydroxycyclohex-1-ene-1-carboxylic acid
Synonyms: shikimate
Molecular Weight (g/mol): 174.15
Molecular Formula: C7H10O5
InChi Key: JXOHGGNKMLTUBP-UHFFFAOYNA-N
SMILES: OC1CC(=CC(O)C1O)C(O)=O
Molecular Weight (g/mol) 
  • (5)
  • (2)
Percent Purity 
  • (3)
  • (2)
  • (2)
Quantity 
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
Physical Form 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (3)
  • (4)
  • (5)
  • (2)
  • (3)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (3)