CAS RN 138-37-4
IUPAC Name:
hydrogen 4-(aminomethyl)benzene-1-sulfonamide chloride
Synonyms:
hydrogen emilene chloride
Molecular Weight (g/mol):
222.69
Molecular Formula:
C7H11ClN2O2S
InChi Key:
SIACJRVYIPXFKS-UHFFFAOYSA-N
SMILES:
[H+].[Cl-].NCC1=CC=C(C=C1)S(N)(=O)=O
1
–
1
of
1
results
1
Homosulfamine Hydrochloride 98.0+%, TCI America™
CAS: 138-37-4 Molecular Formula: C7H11ClN2O2S Molecular Weight (g/mol): 222.687 MDL Number: MFCD00013005 InChI Key: SIACJRVYIPXFKS-UHFFFAOYSA-N Synonym: alpha-Amino-p-toluenesulfonamide Hydrochloride, Mafenide Hydrochloride PubChem CID: 67313 IUPAC Name: 4-(aminomethyl)benzenesulfonamide;hydrochloride SMILES: C1=CC(=CC=C1CN)S(=O)(=O)N.Cl