accessibility menu, dialog, popup

UserName

CAS RN 137897-99-5

IUPAC Name: 1,3-dichloro-5-[(3,5-dichlorophenyl)disulfanyl]benzene
Synonyms: 1,3-dichloro-5-[(3,5-dichlorophenyl)disulfanyl]benzene
Molecular Weight (g/mol): 356.1
Molecular Formula: C12H6Cl4S2
InChi Key: JMQANWHMOHXBEA-UHFFFAOYSA-N
SMILES: ClC1=CC(SSC2=CC(Cl)=CC(Cl)=C2)=CC(Cl)=C1
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results