accessibility menu, dialog, popup

UserName

NaH3COO

CAS RN 137-40-6

IUPAC Name: sodium propanoate
Synonyms: sodium propionate
Molecular Weight (g/mol): 96.06
Molecular Formula: C3H5NaO2
InChi Key: JXKPEJDQGNYQSM-UHFFFAOYSA-M
SMILES: [Na+].CCC([O-])=O
Molecular Weight (g/mol) 
  • (7)
  • (4)
Percent Purity 
  • (2)
  • (4)
  • (2)
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (3)
Physical Form 
  • (5)
Grade 
  • (4)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (9)
  • (4)
  • (4)
  • (2)
  • (5)
  • (7)
  • (4)
  • (2)
  • (4)
  • (2)
  • (3)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (3)
  • (5)
  • (4)