accessibility menu, dialog, popup

UserName

CAS RN 137-04-2

IUPAC Name: hydrogen 2-chloroaniline chloride
Synonyms: hydrogen 2-chloroaniline chloride
Molecular Weight (g/mol): 164.03
Molecular Formula: C6H7Cl2N
InChi Key: DRGIDRZFKRLQTE-UHFFFAOYSA-N
SMILES: [H+].[Cl-].NC1=CC=CC=C1Cl
Percent Purity 
  • (1)
  • (2)
Quantity 
  • (1)
  • (2)
Physical Form 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)